VWR® Socket Caps and Colour Inserts for Screw Microtubes
New Product
Supplier: VWR Collection
VWR® Socket Caps and Colour Inserts for Screw Microtubes
Supplier: VWR Collection
New Product
Certificates
Jump to:
About this item
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
4 Options Available Below
Product Details & Documents
Customisable socket caps with O-ring, accept colourful cap inserts to organise samples or match kit branding. For use with VWR® screw cap microtubes only.
Feature ¾ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Product Family Options
Product Information
- OpisAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product