Order Entry
Poland
Orders LinkContactUsLinkComponent
ML-9 ≥98% protein kinase inhibitor | Avantor
ML-9 ≥98% protein kinase inhibitor
 :   Abcam
ML-9 ≥98% protein kinase inhibitor
 :  Abcam
ML-9 ≥98% protein kinase inhibitor
Broad spectrum ATP-competitive protein kinase inhibitor (Ki values are 32, 4, and 54 μM for PKA, MLCK, and PKC respectively). Ca²+ channel blocker. Shows vasorelaxant effects in vivo.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  1-(5-Chloronaphthalene-1-sulphonyl)-1H-hexahydro-1,4-diazepine hydrochloride, 1-((5-Chloronaphthalen-1-yl)sulphonyl)-1,4-diazepane hydrochloride
  •  :105637-50-1
  •  :361,3 g/mol
  • Constituents::
    100% ML-9
Product Family Options
unology General
  •  
  •    
    1 * 10 mg
    1 * 10 mg - 469 zł
    469 zł
    1 * 10 mg
       469 zł
  •  
  •    
    1 * 50 mg
    1 * 50 mg - 1 810 zł
    1 810 zł
    1 * 50 mg
       1 810 zł
  •    
    1 * 250 mg
    1 * 250 mg - 4 960 zł
    4 960 zł
    1 * 250 mg
       4 960 zł