Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Cambinol ≥98% SIRT1/2 inhibitor

Supplier:   Abcam
Cambinol ≥98% SIRT1/2 inhibitor
Supplier:  Abcam
Certificates

2 Options Available Below

Product Details & Documents

SIRT1/2 inhibitor (IC₅₀ values are 56 and 59 μM at SIRT1 and SIRT2 respectively). Class III HDAC inhibitor, with no inhibition of class I and II. Promotes G₂ cell cycle arrest. Antitumour activity in vitroand in vivo.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  CAmbinol, 5-[(2-Hydroxy-1-naphthyl)methyl]-6-phenyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone, 5-[(2-Hydroxynaphthalen-1-yl)methyl]-6-phenyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, NSC 112546
  • CAS Number:14513-15-6
  • Molecular Weight:360,4 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% 5-[(2-hydroxynaphthalen-1-yl)methyl]-6-phenyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 781 zł
    781 zł
    1 * 5 mg
       781 zł

  • Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 2 950 zł
    2 950 zł
    1 * 25 mg
       2 950 zł