Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Midostaurin ≥98% Broad spectrum protein kinase inhibitor

Supplier:   Abcam
Midostaurin ≥98% Broad spectrum protein kinase inhibitor
Supplier:  Abcam
Certificates
Midostaurin ≥98% Broad spectrum protein kinase inhibitor

2 Options Available Below

Product Details & Documents

Broad spectrum protein kinase inhibitor. Inhibits PKCα, PKCβ and PKCγ isoforms, PDFRβ, VEGFR2, Syk, PKC?, Flk-1, Flt3, Cdk1/B, PKA, c-Kit, c-Fgr, c-Src, VEGFR1 and EGFR. Antitumor activity. Active in vivo.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide
  • CAS Number:120685-11-2
  • Molecular Weight:570,6 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% N-[(2S,3R,4R,6R)-3-methoxy-2-methyl-16-oxo-29-oxa-1,7,17-triaza-octacyclo [12,12,2,12,6,07,28,08,13,015,19,020,27,021,26]nonacosa-8,10,12,14,19,21,23,25,27- nonaen-4-yl]-N-methylbenzamide
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 893 zł
    893 zł
    1 * 1 mg
       893 zł

  • Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 3 510 zł
    3 510 zł
    1 * 5 mg
       3 510 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.