(2-Hydroxylpropyl)-β-cyclodextrine ≥95% modified cyclodextrin
Product Details & Documents
Cyclic oligosaccharide containing seven D-(+)-glucopyranose units that is widely used to improve the aqueous solubility of various compounds, especially those containing a phenyl group. The circular arrangement of its glucose units produces a torus-shaped ring configuration in which the CH₂ groups and ether linkages of the molecule face the hollow interior, resulting in a nonpolar, hydrophobic cavity and a polar, hydrophilic exterior. When combined in solution with other compounds, the nonpolar aromatic portions of that compound interact with the nonpolar interior of the HP-β-CD molecule, thus isolating the aromatic portion of the molecule from the water and thereby increasing its aqueous solubility.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:128446-35-5
Specifications
Specification Test Results
- Constituents::95% 2-Hydroxypropyl-ß-cyclodextrin