Mouse IL-1 beta ELISA kit
Catalog # ABCAAB108866-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IL-1 beta ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse IL-1 beta ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-1 beta in Mouse in plasma, cell culture supernatant, serum samples.