Mouse SCF ELISA Kit
Catalog # ABCAAB100740-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:SCF
- Description:Mouse SCF ELISA kit
- Sample type:Plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:2,74 - 2000 pg/ml
- Storage temperature:4 °C
- Sensitivity:5 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse SCF ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse SCF in Mouse, Rat in plasma, cell culture supernatant, Serum samples.