Thyroxine (Total T4) ELISA Kit
Catalog # ABCAAB178664-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Thyroxine (Total T4) ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Thyroxine (Total T4) ELISA kit is a Competitive ELISA kit for the measurement of Thyroxine (Total T4) in Human in citrate plasma, heparin plasma, serum samples.