Human Von Willebrand Factor ELISA Kit (with plasma controls)
Catalog # ABCAAB168548-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Von Willebrand Factor ELISA Kit (with plasma controls)
- Tests per kit:96 tests
Specifications
About this item
Human Von Willebrand Factor ELISA Kit (with plasma controls) is a Sandwich (quantitative) ELISA kit for the measurement of Human Von Willebrand Factor (with plasma controls) in Human in Plasma, Serum samples.