Order Entry
Poland
ContactUsLinkComponent
JNK (Thr183/Tyr185) In-Cell ELISA Kit
JNK (Thr183/Tyr185) In-Cell ELISA Kit
Catalog # ABCAAB126424-96
Supplier:  Abcam
CAS Number:  
undefined
JNK (Thr183/Tyr185) In-Cell ELISA Kit
Catalog # ABCAAB126424-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    JNK (Thr183/Tyr185) In-Cell ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
JNK (Thr183/Tyr185) In-Cell ELISA Kit is Cell-based (qualitative) ELISA for the measurement of JNK (Thr183/Tyr185) production in Mouse, Rat, Human in Adherent cells samples.