Order Entry
Poland
ContactUsLinkComponent
Mouse FGF-21 ELISA Kit, Fluorescent
Mouse FGF-21 ELISA Kit, Fluorescent
Catalog # ABCAAB229382-1X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse FGF-21 ELISA Kit, Fluorescent
Catalog # ABCAAB229382-1X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse FGF-21 ELISA kit, Fluorescent
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse FGF-21 ELISA kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Mouse FGF-21 with a sensitivity of 1 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Fluorescent Sandwich ELISA - 530/570/590 nm readout: works on any standard plate reader.