Human ICAM1 ELISA Set (without plates)
Catalog # ABCAAB47349-480
Supplier: Abcam
CAS Number:
Specifications
- Description:Human ICAM1 ELISA set (without plates)
- Seq:1
- Storage temperature:+4 ℃
- Tests per kit:5×96 tests
Specifications
About this item
Human ICAM1 ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human ICAM1 ELISA Set (without plates) in Human in plasma, cell culture supernatant, serum samples.