Mouse IP10 ELISA Kit
Catalog # ABCAAB100675-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse IP10 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse IP10 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IP10 in Mouse in Plasma, Cell culture supernatant, Serum samples.