Rat CXCL5 ELISA Kit
Catalog # ABCAAB100775-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat CXCL5 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Rat CXCL5 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat CXCL5 in Rat in Cell culture extracts, Tissue Extracts samples.