Human Perforin ELISA Set (without plates)
Catalog # ABCAAB83709-480
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:Perforin
- Description:Human Perforin ELISA Set (without plates)
- Detection method:Colorimetric
- Time to Results:2 h 45 min
- Detection range:62,5 - 2000 pg/ml
- Storage temperature:4 °C
- Sensitivity:40 pg/ml
- Regulatory status:RUO
- Tests per kit:5×96 tests
Specifications
About this item
Human Perforin ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human Perforin ELISA Set (without plates) in Human in cell culture supernatant, Serum, plasma samples.