Monkey Cystatin C ELISA Kit
Catalog # ABCAAB190542-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Monkey Cystatin C ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Monkey Cystatin C ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Monkey Cystatin C in Monkey in plasma, serum samples.