Bovine Interferon gamma ELISA Kit (IFNG)
ABCAAB193681-96
: Abcam
:
- Description:Bovine Interferon gamma ELISA Kit (IFNG)
- Tests per kit:96 tests
Bovine Interferon gamma ELISA Kit (IFNG) is a Sandwich (quantitative) ELISA kit for the measurement of Bovine Interferon gamma (IFNG) in Cow in Plasma, Cell culture supernatant samples.