Order Entry
Poland
ContactUsLinkComponent
Rat TIMP-2 ELISA Kit
Rat TIMP-2 ELISA Kit
  ABCAAB213923-96
 :  Abcam
 :  
undefined
Rat TIMP-2 ELISA Kit
  ABCAAB213923-96
 :  Abcam
 :  
  • Description:
    Rat TIMP-2 ELISA kit
  • Tests per kit:
    96 tests
Rat TIMP-2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat TIMP-2 in Rat in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum, Tissue Homogenate samples.