Rat PDGF-AA ELISA Kit
Catalog # ABCAAB155464-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat PDGF-AA ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rat PDGF-AA ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat PDGF-AA in Rat in Plasma, Cell culture media, Serum samples.