Human HDL ELISA Kit
Catalog # ABCAAB125961-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Target protein:HDL
- Description:Human HDL ELISA kit
- Sample type:Plasma, milk, serum
- Detection method:Colorimetric
- Time to Results:3 h 0 min
- Detection range:6,25 - 50 µg/ml
- Storage temperature:4 °C
- Sensitivity:1 µg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Human HDL ELISA kit is a Competitive ELISA kit for the measurement of Human HDL in Human in plasma, Milk, Serum samples.