p38 MAPK alpha (Thr180/Tyr182) In-Cell ELISA Kit
Catalog # ABCAAB126425-96
Supplier: Abcam
CAS Number:
Specifications
- Description:p38 MAPK alpha (Thr180/Tyr182) In-Cell ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
p38 MAPK alpha (Thr180/Tyr182) In-cell ELISA kit is cell-based (qualitative) ELISA for the measurement of p38 MAPK alpha (Thr180/Tyr182) production in Mouse, Rat, Human in adherent cells samples.