Human Complement C1r ELISA Kit
Catalog # ABCAAB170245-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Complement C1r ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Complement C1r ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Human Complement C1r in Human in Cerebral Spinal Fluid, Saliva, Urine, plasma, Milk, serum samples.