Mouse Fractalkine ELISA Kit (CX3CL1)
Catalog # ABCAAB100683-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Fractalkine ELISA Kit (CX3CL1)
- Tests per kit:96 tests
Specifications
About this item
Mouse Fractalkine ELISA Kit (CX3CL1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Fractalkine (CX3CL1) in Mouse in Plasma, Cell culture supernatant, Serum samples.