Rat Alpha 1 Acid Glycoprotein / AGP ELISA Kit
Catalog # ABCAAB157729-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Alpha 1 Acid Glycoprotein / AGP ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rat Alpha 1 Acid Glycoprotein / AGP ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Alpha 1 Acid Glycoprotein / AGP in Rat in Plasma, Serum samples.