Rat sE-Selectin ELISA Kit (sCD62E)
ABCAAB171334-96
: Abcam
:
- Description:Rat sE-Selectin ELISA kit (sCD62E)
Rat sE-Selectin ELISA kit (sCD62E) is a Sandwich (quantitative) ELISA kit for the measurement of Rat sE-Selectin (sCD62E) in Rat in cell culture supernatant, Citrate plasma, Heparin plasma, serum samples.