Rat Fractalkine ELISA Kit (CX3CL1)
Catalog # ABCAAB100760-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Fractalkine ELISA kit (CX3CL1)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Rat Fractalkine ELISA kit (CX3CL1) is a Sandwich (quantitative) ELISA kit for the measurement of Rat Fractalkine (CX3CL1) in Rat in plasma, cell culture supernatant, serum samples.