Mouse E-Cadherin ELISA Kit (CDH1)
Catalog # ABCAAB100678-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse E-Cadherin ELISA Kit (CDH1)
- Tests per kit:96 tests
Specifications
About this item
Mouse E-Cadherin ELISA Kit (CDH1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse E-Cadherin (CDH1) in Mouse in Plasma, Cell culture supernatant, Serum samples.