Rabbit IL-21 ELISA Kit
Catalog # ABCAAB273246-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rabbit IL-21 ELISA kit
Specifications
About this item
Rabbit IL-21 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rabbit IL-21 in Rabbit in cell culture supernatant, Citrate plasma, EDTA plasma, serum samples.