Human Luteinizing hormone ELISA Kit (LH)
Catalog # ABCAAB178658-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Luteinizing hormone ELISA kit (LH)
- Tests per kit:96 tests
Specifications
About this item
Human Luteinizing hormone ELISA kit (LH) is a sandwich (quantitative) ELISA kit for the measurement of Human Luteinizing hormone (LH) in Human in Citrate plasma, Heparin Plasma, serum samples.
Certifications: RUO