Mouse TIMP2 ELISA Kit
Catalog # ABCAAB100746-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse TIMP2 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse TIMP2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TIMP2 in Mouse in Plasma, Cell culture supernatant, Serum samples.