Order Entry
Poland
ContactUsLinkComponent
Mouse TCA-3 ELISA Kit (CCL1)
Mouse TCA-3 ELISA Kit (CCL1)
Catalog # ABCAAB155460-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse TCA-3 ELISA Kit (CCL1)
Catalog # ABCAAB155460-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Mouse TCA-3 ELISA kit (CCL1)
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Mouse TCA-3 ELISA kit (CCL1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TCA-3 (CCL1) in Mouse in plasma, cell culture supernatant, serum samples.