Mouse Procollagen III N-terminal propeptide (PIIINP) ELISA Kits
Supplier: Abcam
New Product
Certificates
About this item
Mouse Procollagen III N-terminal propeptide (PIIINP) ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Procollagen III N-terminal propeptide (PIIINP) with a sensitivity of 11,21 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.
Frequently Bought Together