EGFR (Tyr845) In-Cell ELISA Kit
ABCAAB126421-96
: Abcam
:
- Description:EGFR (Tyr845) In-Cell ELISA kit
- Tests per kit:96 tests
EGFR (Tyr845) In-Cell ELISA Kit is Cell-based (qualitative) ELISA for the measurement of EGFR (Tyr845) production in Mouse, Rat, Human in Adherent cells samples.