Phospho-Smad1 (S463/465) ELISA Kit
Catalog # ABCAAB279927-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Phospho-Smad1 (S463/465) ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Phospho-Smad1 (S463/465) ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Phospho-Smad1 (S463/465) in Human in Cell Lysate samples.