Order Entry
Poland
ContactUsLinkComponent
Human MBL ELISA Kit (Mannose-Binding Lectin)
Human MBL ELISA Kit (Mannose-Binding Lectin)
Catalog # ABCAAB193709-96
Supplier:  Abcam
CAS Number:  
undefined
Human MBL ELISA Kit (Mannose-Binding Lectin)
Catalog # ABCAAB193709-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human MBL ELISA kit (Mannose-Binding Lectin)
  • Storage temperature:
    +4 ℃
  • Tests per kit:
    96 tests

Specifications

About this item

Human MBL ELISA kit (Mannose-Binding Lectin) is a Sandwich (quantitative) ELISA kit for the measurement of Human MBL (Mannose-Binding Lectin) in Human in plasma, cell culture supernatant, serum samples.