Rabbit C Reactive Protein (PTX1) ELISA Kit
Catalog # ABCAAB157726-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rabbit C Reactive Protein (PTX1) ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Rabbit C Reactive Protein (PTX1) ELISA kit ab157726 is used to quantify Rabbit C Reactive Protein (PTX1) with a sensitivity of 2,0828 ng/ml. Sandwich ELISA kit.