Mouse C Reactive Protein ELISA Kit (PTX1)
Catalog # ABCAAB157712-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse C Reactive Protein ELISA kit (PTX1)
- Tests per kit:96 tests
Specifications
About this item
Mouse C Reactive Protein ELISA Kit (PTX1) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse C Reactive Protein (PTX1) in Mouse in Urine, Plasma, Serum samples.