Order Entry
Poland
ContactUsLinkComponent
Human Desmoglein 2/DSG2 Antibody Pair - BSA and Azide Free
Human Desmoglein 2/DSG2 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB314796-10X96
Supplier:  Abcam
New Product
CAS Number:  
undefined
Human Desmoglein 2/DSG2 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB314796-10X96
Supplier:  Abcam
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    Desmoglein 2/DSG2
  • Description:
    Human Desmoglein 2/DSG2 antibody pair - BSA and azide free
  • Detection range:
    62,5 - 4000 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Human Desmoglein 2/DSG2 antibody pair - BSA and Azide free.