Order Entry
Poland
ContactUsLinkComponent
Mouse/Rat FGF-21 Antibody Pair - BSA and Azide Free
Mouse/Rat FGF-21 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB241742-10X96
Supplier:  Abcam
CAS Number:  
undefined
Mouse/Rat FGF-21 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB241742-10X96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Format:
    Uncoated
  • Target protein:
    FGF21
  • Description:
    Mouse/Rat FGF-21 antibody pair - BSA and azide free
  • Detection range:
    7,8 - 500 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Tests per kit:
    10×96 tests

Specifications

About this item

Mouse/Rat FGF-21 antibody pair - BSA and Azide free.