Order Entry
Poland
ContactUsLinkComponent
Rat CD62L ELISA Kit (Selectin L)
Rat CD62L ELISA Kit (Selectin L)
Catalog # ABCAAB100776-96
Supplier:  Abcam
CAS Number:  
undefined
Rat CD62L ELISA Kit (Selectin L)
Catalog # ABCAAB100776-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    CD62L
  • Description:
    Rat CD62L ELISA kit (Selectin L)
  • Sample type:
    Plasma, cell culture supernatant, serum
  • Detection method:
    Colorimetric
  • Detection range:
    8,23 - 6000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    6 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat CD62L ELISA kit (Selectin L) is a Sandwich (quantitative) ELISA kit for the measurement of Rat CD62L (Selectin L) in Rat in plasma, cell culture supernatant, Serum samples.