Order Entry
Poland
ContactUsLinkComponent
Anti-Histone H3 (phospho S10) Rabbit Monoclonal Antibody [clone: EPR24060-36] (PE)
Anti-Histone H3 (phospho S10) Rabbit Monoclonal Antibody [clone: EPR24060-36] (PE)
Catalog # ABCAAB317889-100UL
Supplier:  Abcam
New Product
undefined
Anti-Histone H3 (phospho S10) Rabbit Monoclonal Antibody [clone: EPR24060-36] (PE)
Catalog # ABCAAB317889-100UL
Supplier:  Abcam
New Product

Specifications

  • Seq:
    1286
  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

PE Rabbit monoclonal [EPR24060-36] to Histone H3 (phospho S10).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR24060-36
Conjugation: PE
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: