Order Entry
Poland
ContactUsLinkComponent
Anti-CD134/OX40L receptor Rabbit Monoclonal Antibody [clone: EPR23000-42] (Alexa Fluor® 750)
Anti-CD134/OX40L receptor Rabbit Monoclonal Antibody [clone: EPR23000-42] (Alexa Fluor® 750)
Catalog # ABCAAB321523-100UL
Supplier:  Abcam
New Product
undefined
Anti-CD134/OX40L receptor Rabbit Monoclonal Antibody [clone: EPR23000-42] (Alexa Fluor® 750)
Catalog # ABCAAB321523-100UL
Supplier:  Abcam
New Product

Specifications

  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 750 Rabbit monoclonal [EPR23000-42] to CD134/OX40L receptor.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD134 / OX40L receptor
Clonality: Monoclonal
Clone: EPR23000-42
Conjugation: Alexa Fluor® 750
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: