Anti-Syntenin Rabbit Monoclonal Antibody [clone: EPR8102]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR8102] to Syntenin - BSA and Azide free.
Type: Primary
Antigen: Syntenin
Clonality: Monoclonal
Clone: EPR8102
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together