Anti-HGF Rabbit Monoclonal Antibody [clone: EPR22870-13]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [EPR22870-13] to HGF - BSA and Azide free (Detector).
Type: Primary
Antigen: HGF
Clonality: Monoclonal
Clone: EPR22870-13
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together