Anti-MAVS Rabbit Monoclonal Antibody [clone: EPR26357-91]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR26357-91] to MAVS.
Type: Primary
Antigen: MAVS
Clonality: Monoclonal
Clone: EPR26357-91
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together