Order Entry
Poland
ContactUsLinkComponent
Anti-smooth muscle Myosin heavy chain 11 Rabbit Monoclonal Antibody [clone: EPR5336(B)] (Alexa Fluor® 488)
Anti-smooth muscle Myosin heavy chain 11 Rabbit Monoclonal Antibody [clone: EPR5336(B)] (Alexa Fluor® 488)
Catalog # ABCAAB197524-100
Supplier:  Abcam
undefined
Anti-smooth muscle Myosin heavy chain 11 Rabbit Monoclonal Antibody [clone: EPR5336(B)] (Alexa Fluor® 488)
Catalog # ABCAAB197524-100
Supplier:  Abcam

Specifications

  • Seq:
    1286
  • Size:
    100 µl
  • Pack type:
    Vial

Specifications

About this item

Alexa Fluor® 488 Rabbit monoclonal [EPR5336(B)] to smooth muscle Myosin heavy chain 11.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: smooth muscle Myosin heavy chain 11
Clonality: Monoclonal
Clone: EPR5336(B)
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human