Order Entry
Poland
ContactUsLinkComponent
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EPR20132-63]
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EPR20132-63]
Supplier:  Abcam
Certificates
undefined
Anti-Antithrombin III/ATIII Rabbit Monoclonal Antibody [clone: EPR20132-63]
Supplier:  Abcam

Specifications

  • Catalog No:
  • ABCAAB244918-1
  • ABCAAB244918-100
  • Antigen symbol:
  • Antithrombin III/ATIII
  • Antithrombin III/ATIII
  • Antigen name:
  • AT3
  • AT3
  • Antigen synonyms:
  • PRO0309|SERPINC1|Antithrombin-III|ATIII|Serpin C1
  • PRO0309|SERPINC1|Antithrombin-III|ATIII|Serpin C1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • EPR20132-63
  • EPR20132-63
  • Form:
  • Liquid
  • Liquid

Specifications

Frequently Bought Together