Anti-Ferredoxin--NADP reductase, apicoplast Rabbit Polyclonal Antibody
Catalog # ABCAAB243175-400
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:PF3D7_0623200
- Antigen symbol:Ferredoxin--NADP reductase, apicoplast
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:FNR
- Molecular weight:43779 Da
- Storage temperature:Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term. Avoid freeze / thaw cycle.
- Concentration:4 mg/ml
- Shipping temperature:4 °C
- Immunogen:Recombinant Full Length Protein corresponding to Plasmodium falciparum 3D7 FNR.
- Purification:Protein A Affinity purification
- Size:400 µg
- Pack type:Vial
Specifications
About this item
Rabbit polyclonal to Ferredoxin--NADP reductase, apicoplast.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Ferredoxin--NADP reductase, apicoplast
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Corn;P. falciparum