Anti-Neuropeptide Y Rabbit Monoclonal Antibody [clone: EPR21877]
Supplier: Abcam
New Product
Certificates
About this item
Rabbit monoclonal [EPR21877] to Neuropeptide Y.
Type: Primary
Antigen: Neuropeptide Y
Clonality: Monoclonal
Clone: EPR21877
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together