Anti-gamma Tubulin Mouse Monoclonal Antibody [clone: GTU-88]
Catalog # ABCAAB11316-100UG
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Tubulin gamma-2 chain
- Antigen symbol:gamma Tubulin
- Clonality:Monoclonal
- Clone:GTU-88
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Seq:1286
- Form:Liquid
- Antigen synonyms:Gamma-2-tubulin|TUBG2
- Molecular weight:51092 Da
- Storage temperature:Upon delivery aliquot and store at –20 °C or –80 °C. Avoid repeated freeze / thaw cycles.
- Shipping temperature:4 °C
- Immunogen:Synthetic Peptide within Human TUBG2 aa 1-100 conjugated to Keyhole Limpet Haemocyanin. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:ICC/IF
- Purification:Proprietary technique
- Size:100 µg
- Pack type:Vial
Specifications
About this item
Mouse monoclonal [GTU-88] to gamma Tubulin - Centrosome Marker.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: gamma Tubulin
Clonality: Monoclonal
Clone: GTU-88
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse; Chicken; Bovine; Dog; Human; African Green Monkey; Chinese hamster